Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥95.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
2-8°C
Quality Segment
type
for DNA synthesis
product line
Proligo Reagents
assay
≥95.0% (Nucleoside Purity, reversed phase HPLC)
pore size
1000 Å
capacity
≥99 % (Coupling Efficiency)
λ
conforms (UV/VIS Identity)
storage temp.
2-8°C
Still not finding the right product?
Explore all of our products under DMT-dC(bz)-CPG
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.