Skip to Content
Merck

C3545

Monoclonal Anti-Calmodulin antibody produced in mouse

clone 6D4, ascites fluid

Synonym(s):

Anti-CALM3, Anti-calmodulin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44
MDL number:
Conjugate:
unconjugated
Clone:
6D4, monoclonal
Application:
WB
Citations:
13
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6D4, monoclonal

contains

15 mM sodium azide as preservative

species reactivity

chicken, Dictyostelium discoideum, bovine, rat

technique(s)

western blot: 1:2,000 using a bovine brain calmodulin preparation

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Calm1(24242)

General description

Calmodulin belongs to a family of structurally homologous Ca2+-binding proteins. CALM1 gene is located on human chromosome 14q32.11.
Calmodulin is a highly conserved 17kDa calcium binding protein found in all eukaryotic cells. It is a multifunctional molecule. Monoclonal anti-Calmodulin (mouse IgG1 isotype) is derived from the 6D4 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with calmodulin, purified from Dictyostelium discoideum and conjugated to KLH.

Immunogen

preparation of purified calmodulin from Dictyostelium discoideum conjugated to KLH.

Application

Monoclonal Anti-Calmodulin antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoblot
  • dot blot
  • immunocytochemistry
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

Calmodulin is a ubiquitous molecule which can bind up to 4 calcium ions, regulating and mediating a wide variety of biochemical processes.
The antibody recognizes native and SDS-denatured calmodulin.
These antibodies are directed against different epitopes within the calmodulin molecule.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
C3545-.2ML04061838104281