Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C10H18N2O6 · C2F3O2H
CAS Number:
Molecular Weight:
376.28
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Quality Level
assay
≥95% (TLC)
form
powder
color
white
storage temp.
2-8°C
SMILES string
OC(=O)C(F)(F)F.NCCCC[C@H](N(CC(O)=O)CC(O)=O)C(O)=O
InChI
1S/C10H18N2O6.C2HF3O2/c11-4-2-1-3-7(10(17)18)12(5-8(13)14)6-9(15)16;3-2(4,5)1(6)7/h7H,1-6,11H2,(H,13,14)(H,15,16)(H,17,18);(H,6,7)/t7-;/m0./s1
InChI key
PKZCFUDHJJGMPN-FJXQXJEOSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hochuli, E., et al.
Journal of Chromatographic Science, 411, 177-177 (1987)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C3205-250MG | 04061832422985 |
| C3205-50MG | 04061832422992 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service