Skip to Content
Merck

C2526

L-Cystine dihydrochloride

≥98% (TLC), suitable for cell culture

Synonym(s):

Cystine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2 · 2HCl
CAS Number:
Molecular Weight:
313.22
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
250-391-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Cystine dihydrochloride, ≥98% (TLC)

Quality Segment

assay

≥98% (TLC)

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

color

white to off-white

mp

≥211.36 °C

SMILES string

Cl.Cl.N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2.2ClH/c7-3(5(9)10)1-13-14-2-4(8)6(11)12;;/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12);2*1H/t3-,4-;;/m0../s1

InChI key

HHGZUQPEIHGQST-RGVONZFCSA-N

Biochem/physiol Actions

L-Cystine is a natural proteinogenic amino acid sulfide dimer of L-cysteine. L-cystine is used as a cell culture supplement. L-cystine is also used to characterize and identify specific amino acid transport systems in cells such as system system Xc(─) .

Disclaimer

hygroscopic; handle in <30% humidity


Still not finding the right product?

Explore all of our products under L-Cystine dihydrochloride


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library