Skip to Content
Merck

C2485

β-Cyclodextrin polymer

powder

Synonym(s):

Beta-Cyclodextrin Polymeric Adsorbent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

form

powder

mol wt

average mol wt 2,000-300,000 Da

composition

Cyclodextrin content, 60% iodometric

color

white

solubility

H2O: soluble 100mg plus 1 mL, clear, colorless to light yellow

storage temp.

2-8°C

General description

Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via α(1,4) glycosidic bonds. Common cyclodextrins contain six (α-CD); seven (β-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivatized through esterification at positions two, three and/or six.

Application

Beta-cyclodextrin (β-Cyclodextrin) may be used in the development of biocompatible drug delivery vehicles and membrane constructs to improve compound solubility in aqueous environments. β-cyclodextrin polymer and its ester derivatives are used for the development and construction of biomaterials such as hydrogels, nanoassemblies and biomatricies. β-cyclodextrin may be used for the development of biosensors.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library