Skip to Content
Merck

C2235

CB 1954

solid

Synonym(s):

5-(1-Aziridinyl)-2,4-dinitrobenzamide, 5-(Aziridin-1-yl)-2,4-dinitrobenzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥94% (HPLC)

Quality Segment

form

solid

storage temp.

−20°C

SMILES string

NC(=O)c1cc(N2CC2)c(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H8N4O5/c10-9(14)5-3-7(11-1-2-11)8(13(17)18)4-6(5)12(15)16/h3-4H,1-2H2,(H2,10,14)

InChI key

WOCXQMCIOTUMJV-UHFFFAOYSA-N

Application

CB 1954 has been used in drug-induced ablation studies.

Biochem/physiol Actions

CB 1954 is an anticancer prodrug used in gene therapy research; activated by NAD(P)H quinone oxidoreductase 2.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library