Skip to Content

C211800

DMT-2′O-MOE-rMeC(bz) Phosphoramidite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H60N5O10P
CAS Number:
Molecular Weight:
922.01
UNSPSC Code:
41116105
Assay:
≥99% (31P-NMR), ≥99.0% (reversed phase HPLC)
Biological source:
non-animal source (no BSE/TSE risk)
Form:
powder
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source (no BSE/TSE risk)

Quality Segment

product line

Proligo Reagents

assay

≥99% (31P-NMR), ≥99.0% (reversed phase HPLC)

form

powder

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC), ≤0.3 wt. % water content (Karl Fischer), ≤0.5% CET-H-Phosphonate (reversed phase HPLC), ≤0.5% DIPA-H-Phosphonate (reversed phase HPLC), ≤0.5% DMT-rMeC(bz)moe (reversed phase HPLC), ≤0.5% DMT-rMeC(bz)moe -amidate (reversed phase HPLC), ≤0.5% DMT-rMeC(bz)moe-Bz (reversed phase HPLC), ≤0.5% H-Phosphonat (reversed phase HPLC), ≤0.5% O3, O5-Bis-DMT-rMeC(bz)moe (reversed phase HPLC), ≤0.5% P(III) 100-169ppm (31P-NMR), ≤3 wt. % residual solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR, conforms to structure for LC-MS

storage temp.

−20°C



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library