C2105
Z-Val-Ala-Asp fluoromethyl ketone
powder, ≥90% (TLC)
Synonym(s):
Z-VAD-FMK, Non-methylated
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H28FN3O7
CAS Number:
Molecular Weight:
453.46
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥90% (TLC)
form
powder
shipped in
dry ice
storage temp.
−20°C
SMILES string
CC(C)[C@H](NC(=O)OCc1ccccc1)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(O)=O)C(=O)CF
InChI
1S/C21H28FN3O7/c1-12(2)18(25-21(31)32-11-14-7-5-4-6-8-14)20(30)23-13(3)19(29)24-15(9-17(27)28)16(26)10-22/h4-8,12-13,15,18H,9-11H2,1-3H3,(H,23,30)(H,24,29)(H,25,31)(H,27,28)/t13-,15-,18-/m0/s1
InChI key
SUUHZYLYARUNIA-YEWWUXTCSA-N
Amino Acid Sequence
Z-Val-Ala-Asp-OMe-FMK
Biochem/physiol Actions
Non-methylated competitive and irreversible inhibitor of caspase-1, as well as other caspases, which can be used directly with purified enzymes. It does not require esterase to hydrolyze the O-methyl ester like the cell-permeable form, Z-Val-Ala-Asp(OMe) fluoromethyl ketone (Product Code V-116).
Features and Benefits
This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Longlong Xie et al.
Signal transduction and targeted therapy, 5(1), 56-56 (2020-05-21)
Latent membrane protein 1 (LMP1) is a major Epstein-Barr virus (EBV)-encoded oncoprotein involved in latency infection that regulates mitochondrial functions to facilitate cell survival. Recently, mitochondrial fission has been demonstrated as a crucial mechanism in oncovirus-mediated carcinogenesis. Mitochondrial dynamin-related protein
Xintong Jiang et al.
JACC. Basic to translational science, 4(3), 304-317 (2019-07-18)
CANTOS (Canakinumab Antiinflammatory Thrombosis Outcome Study) confirmed interleukin (IL)-1β as an appealing therapeutic target for human atherosclerosis and related complications. However, there are serious gaps in our understanding of IL-1 production in atherosclerosis. Herein the authors show that complex plaques
Teruyuki Kajiume et al.
Cell death discovery, 4, 111-111 (2018-12-12)
Mature neutrophils must be quickly removed from inflammatory sites to prevent tissue damage. Neutrophil removal is thought to be accomplished primarily through caspase-dependent apoptosis, which involves several genes of mitochondrial origin. However, mature neutrophils show reduced gene transcription and mitochondrial
Tonghui Li et al.
Frontiers in pharmacology, 13, 854526-854526 (2022-06-07)
Aloe-emodin (1,8-dihydroxy-3-hydroxymethyl-anthraquinone), derived from some Chinese edible medicinal herbs, exerts a potential anticancer activity on various cancer cells, making it a drug candidate for cancer therapy. Yet, the role of aloe-emodin in pyroptosis, a new type of cell death, is
Feng Shi et al.
Theranostics, 9(9), 2424-2438 (2019-05-28)
EBV infection is a recognized epigenetic driver of carcinogenesis. We previously showed that EBV could protect cancer cells from TNF-induced necroptosis. This study aims to explore the epigenetic mechanisms allowing cancer cells with EBV infection to escape from RIP3-dependent necroptosis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service