Merck
C1420
Cuvette washer
Double
Synonym(s):
cuvette cleaning
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
SB.32
UNSPSC Code:
41121813
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
General description
Supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvettes.
Still not finding the right product?
Explore all of our products under Cuvette washer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
C1420-1EA04061826185827