Skip to Content
Merck
All Photos(2)

Key Documents

C1335

Sigma-Aldrich

S-(−)-Carbidopa

≥98% (TLC), powder

Synonym(s):

(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic acid, S-(−)-α-Hydrazino-3,4-dihydroxy-2-methylbenzenepropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O4
CAS Number:
Molecular Weight:
226.23
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

methanol and 1 drop HCl: 4.90-5.10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(Cc1ccc(O)c(O)c1)(NN)C(O)=O

InChI

1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)/t10-/m0/s1

InChI key

TZFNLOMSOLWIDK-JTQLQIEISA-N

Gene Information

human ... DDC(1644)

Looking for similar products? Visit Product Comparison Guide

General description

Carbidopa is used for treating restless legs (RL) syndrome and periodic leg movements in sleep (PLMS). Carbidopa along with levodopa is used to treat Parkinson′s disease.

Biochem/physiol Actions

Peripheral inhibitor of L-aromatic amino acid decarboxylase.
Peripheral inhibitor of L-aromatic amino acid decarboxylase. When co-administered with L-DOPA, it prevents extra-CNS conversion to dopamine, thereby increasing CNS concentrations of effective compounds. Selectively cytotoxic to human pulmonary carcinoid and small cell lung carcinoma cells by increasing H2O2 in these cell lines, which are sensitive to reactive oxygen species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Deleu et al.
Naunyn-Schmiedeberg's archives of pharmacology, 348(6), 576-581 (1993-12-01)
The effect of carbidopa on the pharmacokinetics and metabolism of levodopa (L-dopa) in blood plasma and skeletal muscle extracellular fluid (ECF) has been studied by repeated measurements in one beagle dog. The administration of a single dose of L-dopa (25
S Kaakkola et al.
Journal of neurochemistry, 60(1), 137-144 (1993-01-01)
The effects of two new catechol-O-methyltransferase (COMT) inhibitors, OR-611 and Ro 40-7592, in combination with L-3,4-dihydroxyphenylalanine (L-dopa) with or without carbidopa on extracellular levels of dopamine (DA), 3,4-dihydroxyphenylacetic acid (DOPAC), homovanillic acid (HVA), 3-O-methyldopa (3-OMD), and 5-hydroxyindoleacetic acid in rat
Levodopa with benserazide or carbidopa in Parkinson disease
Rinne UK and Molsa P
Neurology, 29(12), 1584-1589 (1979)
Pergolide and carbidopa/levodopa treatment of the restless legs syndrome and periodic leg movements in sleep in a consecutive series of patients
Earley CJ and Allen RP
Sleep, 19(10), 801-810 (1996)
Severe gastrointestinal complications in patients with levodopa/carbidopa intestinal gel infusion.
Fabian Klostermann et al.
Movement disorders : official journal of the Movement Disorder Society, 27(13), 1704-1705 (2012-11-30)

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service