Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human blood (monocyte)
Morphology:
(macrophages)
biological source
human blood (monocyte)
product line
PromoCell®
packaging
pkg of 500,000 cells
morphology
(macrophages)
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Application
Cryopreserved human monocyte-derived macrophages (hMDM) are convenient and easy-to-handle. The thawed cells plate into all tissue culture vessel formats and can be maintained as adherent, biologically functional cultures for several weeks. Optionally, user-customizable activation of the cells can be performed.Cryopreserved human macrophages are produced from monocytes in M2 Macrophage Generation Media DXF and are available as non-activated, fully qualified M2- (M-CSF) polarized cells. After 9-10 days of differentiation, the M2 macrophages are cryopreserved using Cryo-SFM. Each cryo vial contains more than 5 million viable cells after thawing.
Biochem/physiol Actions
Monocyte
Preparation Note
Click here for more information.
Other Notes
Recommended Plating Density: 200000 cells per cm2Tested Markers: CD163 positive, CD68 positiveNote: The plated cells do not proliferate. Recommended plating density for obtaining an adherent cell monolayer with 70-90% confluency.
Recommended Primary Cell Culture Media:Link
Recommended Primary Cell Culture Media:Link
Legal Information
PromoCell is a registered trademark of PromoCell GmbH
Disclaimer
Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Still not finding the right product?
Explore all of our products under Human M2 Macrophages (hMDM-MCSF(-))
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.