Skip to Content

C-12914

Human M1 Macrophages (hMDM-GMCSF(-))

Monocyte-derived, single donor, 1.5 million cryopreserved cells

Synonym(s):

hMDM-GMCSF(-) cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human blood (monocyte)
Morphology:
(macrophages)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human blood (monocyte)

product line

PromoCell®

packaging

pkg of 1,500,000 cells

morphology

(macrophages)

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Application

Cryopreserved human monocyte-derived macrophages (hMDM) are convenient and easy-to-handle. The thawed cells plate into all tissue culture vessel formats and can be maintained as adherent, biologically functional cultures for several weeks. Optionally, user-customizable activation of the cells can be performed.Cryopreserved human macrophages are produced from monocytes in M1 Macrophage Generation Media DXF and are available as non-activated, fully qualified M1- (GM-CSF) polarized cells. After 9-10 days of differentiation, the M1 macrophages are cryopreserved using Cryo-SFM. Each cryo vial contains more than 1.5 million viable cells after thawing.

Biochem/physiol Actions

Monocyte

Preparation Note

Click here for more information.

Other Notes

Recommended Plating Density: 100000 cells per cm2Tested Markers: CD80 positive, CD68 positiveNote: The plated cells do not proliferate. Recommended plating density for obtaining an adherent cell monolayer with 70-90% confluency.
Recommended Primary Cell Culture Media:Link
Recommended Primary Cell Culture Media:Link

Legal Information

PromoCell is a registered trademark of PromoCell GmbH

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library