Skip to Content
Merck

C1241

Anti-Calcium Channel CaV1.2 (human) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Human α1C, L-Type of Voltage-gated Calcium Channel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable, western blot: 1:200 using rat heart membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNA1C(775)
mouse ... Cacna1c(12288)
rat ... Cacna1c(24239)

General description

Calcium Channel CaV1.2 (CACNA1C ) encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization.

Immunogen

peptide corresponding to residues 2-15 of human Cav1.2 (exon 1B). The sequence has 14/15 residues identical in mouse and rat.

Biochem/physiol Actions

Calcium channel, voltage-dependent, L type, α1C subunit (Cav1.2) is a protein encoded by the CACNA1C gene in humans and belongs to the family of voltage-gated calcium channels. It serves as the key transducers of cell surface membrane potential changes into local intracellular calcium transients that initiate different physiological events. CACNA1C is a subunit of L-type voltage-dependent calcium channel. It is mapped to chromosome 12p13.3 and is implicated as a susceptibility gene for schizophrenia. CACNA1C is selectively expressed in human TH2 cells and blockage of Cav1.2 channel activation in TH2 cells may represent new strategies to treat allergic diseases in human subjects. Its increased activity is implicated in the pathogenesis of dementia and Alzheimer′s disease (AD).
Recognizes mouse Cav1.2, splice variants P22002-4 and P22002-5 of rat Cav1.2 and splice variant P15381-4 of rabbit Cav1.2.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, with 1% BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library