Skip to Content
Merck

C0897

Citrate Lyase from Klebsiella pneumoniae

lyophilized powder, ≥0.20 unit/mg solid

Synonym(s):

Citrate oxaloacetate lyase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-740-7
MDL number:
EC Number:
Specific activity:
≥0.20 unit/mg solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized powder

Quality Segment

specific activity

≥0.20 unit/mg solid

mol wt

>5 kDa

storage temp.

2-8°C

Application

Citrate Lyase from Klebsiella pneumoniae has been used in the digestion of low molecular weight human milk fraction (>5 kDa referred to as 5kF).

Biochem/physiol Actions

Citrate lyase is found in several microorganisms and catalyzes the first step of citrate degradation, forming acetate and oxaloacetate. The enzyme contains 3 polypeptide subunits, α-subunit (a transferase), β-subunit (acyl lyase) and γ-subunit (acyl-carrier protein).

Physical form

Lyophilized powder containing bovine serum albumin, sucrose, and MgSO4

Other Notes

One unit will convert 1.0 μmole of citrate to oxalacetate per min at pH 7.6 at 25 °C.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library