Key Documents
BMSIPS2050PS4
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
4-arm, stainless steel
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-S4
diam. × L
50 mm × 400 mm
General description
4-arm stainless steel propeller is useful for mid to high speed mixing.
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Application
stirring, mixing