Skip to Content
Merck
All Photos(2)

Key Documents

B9061

Sigma-Aldrich

Bicalutamide (CDX)

≥98% (HPLC), powder

Synonym(s):

Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

solubility

DMSO: >5 mg/mL
H2O: insoluble

originator

AstraZeneca

SMILES string

CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F

InChI

1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)

InChI key

LKJPYSCBVHEWIU-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

Application

Bicalutamide (CDX) has been used as an androgen receptor (AR) antagonist in prostate, bladder cancer cell lines and human fetal skeletal muscle cells. It has also been used as a supplement in RPMI 1640 for culturing androgen-independent LNCaP (LNCaP-AI) cell line.

Biochem/physiol Actions

Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) Antagonist and a pure antiandrogen.
Bicalutamide (CDX) is a non-steriodal Androgen Receptor (AR) antagonist and a pure antiandrogen. It acts via balancing histone acetylation/deacetylation and recruitment of coregulators. Bicalutamide (CDX) abolishes androgen-mediated expression. For example, MMP13 upregulation in prostate cancer, PLZF (promyelocytic leukemia zinc finger protein), and GADD45γ (growth arrest and DNA damage inducible, gamma). Bicalutamide (CDX) is inhibited by non-genomic, transcription-independent stimulation of PI3K/AKT phosphorylation by androgens.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive stroma component COL6A1 is upregulated in castration-resistant prostate cancer and promotes tumor growth
Zhu YP, et al.
Testing, 6(16), 14488-14488 (2015)
S Belikov et al.
Molecular and cellular endocrinology, 365(1), 95-107 (2012-10-16)
Prostate cancer growth depends on androgens. Synthetic antiandrogens are used in the cancer treatment. However, antiandrogens, such as bicalutamide (BIC), have a mixed agonist/antagonist activity. Here we compare the antiandrogenic capacity of BIC to a new antiandrogen, MDV3100 (MDV) or
Maha Hussain et al.
The New England journal of medicine, 368(14), 1314-1325 (2013-04-05)
Castration resistance occurs in most patients with metastatic hormone-sensitive prostate cancer who are receiving androgen-deprivation therapy. Replacing androgens before progression of the disease is hypothesized to prolong androgen dependence. Men with newly diagnosed, metastatic, hormone-sensitive prostate cancer, a performance status
Testosterone insulin-like effects: an in vitro study on the short-term metabolic effects of testosterone in human skeletal muscle cells
Antinozzi C, et al.
Journal of Endocrinological Investigation, 40(10), 1133-1143 (2017)
Androgen receptor antagonist bicalutamide induces autophagy and apoptosis via ULK2 upregulation in human bladder cancer cells
Hao K, et al.
International Journal of Clinical and Experimental Pathology, 10(7), 7603-7615 (2017)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service