Skip to Content
Merck

B7554

Anti-c-Myc−Biotin antibody, Mouse monoclonal

clone 9E10, purified from hybridoma cell culture

Synonym(s):

Anti-c-Myc, Monoclonal Anti-c-Myc−Biotin antibody produced in mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.56
UNSPSC Code:
12352203
Conjugate:
biotin conjugate
Clone:
9E10, monoclonal
Application:
WB CL
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

biotin conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

9E10, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

0.8 mg/mL

technique(s)

western blot (chemiluminescent): 0.05-0.1 μg/mL using an extract of 293T cells or E. coli cells expressing c-Myc tagged fusion protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

The Monoclonal Anti-c-Myc mouse antibody is isolated from ascites and conjugated to biotin. The antibody recognizes the c-Myc tag sequence on c-Myc tagged fusion proteins when expressed N- or C terminal to the fusion protein.

Immunogen

synthetic peptide of the human p62c-Myc protein.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
The antibody is suitable for immunoblotting.

Biochem/physiol Actions

Staining by immunoblotting is specifically inhibited by the c-Myc peptide (Product Code M2435).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide as a preservative.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library