Skip to Content
Merck
All Photos(1)

Key Documents

B6766

Sigma-Aldrich

BME Amino Acids Solution 50×

Without L-glutamine, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Amino Acids Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

solution

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

ambient

storage temp.

2-8°C

Application

BME amino acids solution 50X has been used for embryo culture.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination of oviduct fluid and heparin to improve monospermic zygotes production during porcine in vitro fertilization
Batista RITP, et al.
Theriogenology, 86(2), 495-502 (2016)
Influence of sperm fertilising concentration, sperm selection method and sperm capacitation procedure on the incidence of numerical chromosomal abnormalities in IVF early bovine embryos
Demyda-Peyras S, et al.
Reproduction, Fertility, and Development, 27(2), 351-359 (2015)
Different co-culture systems have the same impact on bovine embryo transcriptome
Carvalho AV, et al.
Reproduction (Cambridge, England), 154(5), 695-710 (2017)
Tianbin Liu et al.
Cellular reprogramming, 22(6), 282-290 (2020-11-13)
Essential amino acids (EAA) of inappropriate concentration have been reported to compromise the development of embryo. This study aimed to investigate the effect of EAA on the developmental competence of porcine embryos produced by either handmade cloning (HMC) or parthenogenetic
Hongli Li et al.
In vitro cellular & developmental biology. Animal, 55(8), 586-597 (2019-08-02)
Rotavirus (RV) is the leading cause of viral gastroenteritis in neonates and VP6 protein has been discussed as a potential candidate vaccine. CRISPR/Cas9 was the latest generation of gene editing tools that can mediate the site-specific knock-in of exogenous genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service