Skip to Content
Merck
All Photos(1)

Key Documents

B6024

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

tablet, suitable for identification of lac+ bacterial colonies

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1402009
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, tablet

form

tablet

suitability

suitable for identification of lac+ bacterial colonies

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Bromo-4-chloro-3-indolyl β -D-galactopyranoside, commonly known as X-Gal, is a histochemical substrate for β-galactosidase.

Application

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside has been used in the preparation of X-Gal stock solution to add it in LB agar plates for blue-white screening of transformants.
X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Biochem/physiol Actions

X-gal is routinely used to detect transgene activity in situ.

Quantity

Contains 5 mg substrate per tablet

Reconstitution

A stock solution (100 mg/ml) is prepared by dissolving X-Gal in dimethylformamide and should be stored at −20°C and protected from light. Sterilization is not required.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Creation of recombinant antigen-binding molecules derived from hybridomas secreting specific antibodies
Fields C, et al.
Nature Protocols, 8(6), 1125-1125 (2013)
Analysis of Transgene Expression
Transactions of the American Clinical and Climatological Association, 543-564 (2014)
Hiroyuki Kanazawa et al.
PloS one, 6(4), e19195-e19195 (2011-05-12)
Ischemia-reperfusion (I/R) injury associated with living donor liver transplantation impairs liver graft regeneration. Mesenchymal stem cells (MSCs) are potential cell therapeutic targets for liver disease. In this study, we demonstrate the impact of MSCs against hepatic I/R injury and hepatectomy.
Yibing Wang et al.
PloS one, 8(3), e59195-e59195 (2013-03-26)
Our study had three objectives: to extend the plasmid-based transformation protocol to a clinical isolate of C. trachomatis belonging to the trachoma biovar, to provide "proof of principle" that it is possible to "knock out" selected plasmid genes (retaining a
Enzo R Porrello et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 187-192 (2012-12-19)
We recently identified a brief time period during postnatal development when the mammalian heart retains significant regenerative potential after amputation of the ventricular apex. However, one major unresolved question is whether the neonatal mouse heart can also regenerate in response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service