Skip to Content
Merck
All Photos(2)

Key Documents

B5144

Sigma-Aldrich

Bromelain from pineapple stem

≥4 units/mg protein, (chromatography purified)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

(chromatography purified)
≥4 units/mg protein

composition

Protein, 30-50% biuret

solubility

H2O: soluble 1.0 mg/mL, clear, colorless

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bromelain from pineapple stem has been used as a positive control in sodium dodecyl sulfate (SDS) / polyacrylamide gel electrophoresis (PAGE) zymograms. It has also been used as a Azocasein substrate for the determination of proteolytic activity.
Bromelain may be used to inhibit the biosysnthesis of proinflammatory prostaglandins. It may be used to reduce clotting efficiency. Bromelain, from pineapple stem, has been used to make enzymatic hydrolysates of honeybee-collected pollen .

Biochem/physiol Actions

Bromelain is a cysteine endopeptidase with broad specificity for cleavage of proteins. Bromelain may be from a stem or piece of fruit. Product P3169 (pGlu-Phe-Leu-p-nitroanilide) may be used as a chromogenic substrate for the assay of bromelain.

Unit Definition

One unit will release 1.0 μmole of p-nitrophenol from N-α-Z-L-lysine p-nitrophenyl ester per min at pH 4.6 at 25 °C.

Physical form

Lyophilized powder containing mannitol and potassium phosphate buffer salts

Preparation Note

Chromatographically purified
Chromatographically purified. Dissolves in water at 1 mg/mL concentration to form a clear, colorless solution.

inhibitor

Product No.
Description
Pricing

substrate

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azocasein substrate for determination of proteolytic activity: reexamining a traditional method using bromelain samples
Coelho DF, et al.
BioMed Research International (2016)
Ashley G Yow et al.
Scientific reports, 13(1), 11605-11605 (2023-07-19)
Papain (aka C1A) family proteases, including bromelain enzymes, are widespread across the plant kingdom and play critical regulatory functions in protein turnover during development. The proteolytic activity exhibited by papain family proteases has led to their increased usage for a
Larisa V Kordyukova et al.
Membranes, 11(10) (2021-10-23)
Influenza A virus envelope contains lipid molecules of the host cell and three integral viral proteins: major hemagglutinin, neuraminidase, and minor M2 protein. Membrane-associated M1 matrix protein is thought to interact with the lipid bilayer and cytoplasmic domains of integral
Acidic cysteine protease inhibitors from pineapple stem.
R L Heinrikson et al.
Methods in enzymology, 45, 740-751 (1976-01-01)
Larisa Kordyukova et al.
Methods in molecular biology (Clifton, N.J.), 1934, 265-291 (2019-07-01)
Palmitoylation or S-acylation is the posttranslational attachment of fatty acids to cysteine residues and is common among integral and peripheral membrane proteins. Palmitoylated proteins have been found in every eukaryotic cell type examined (yeast, insect, and vertebrate cells), as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service