Skip to Content
Merck

B5020

Buffer, reference standard

pH 4.00±0.01 (25 °C)

Synonym(s):

pH calibration solution at 25 °C pH 4.00±0.01

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Level

color

color-coded red

pH

4.00±0.01 (25 °C)

storage temp.

15-25°C

Application

The buffer reference standard was used in the calibration of a pH probe and pH meter in analytical procedures.

Features and Benefits

  • Traceable to NIST standard reference materials
  • A ready-to-use solution available in various packaging to provide ease-of-use

Packaging

500 mL size packaged in polyethylene bottle; 1 gal size in convenient shelf-top dispenser with spigot.

Analysis Note

Standardized against N.I.S.T. standard reference materials.


Still not finding the right product?

Explore all of our products under Buffer, reference standard


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



An integrated flex-microfluidic-Si chip device towards sweat sensing applications
Nie C, et al.
Sensors and Actuators B, Chemical, 227, 427-437 (2016)
Hydrochloric acid alters the effect of L-glutamic acid on cell viability in human neuroblastoma cell cultures
Croce N, et al.
Journal of Neuroscience Methods, 217(1-2), 26-30 (2013)



Global Trade Item Number

SKUGTIN
B5020-6X500ML04061833433560
B5020-500ML04061837901553
B5020-1GA04061837901546