Skip to Content
Merck

B3681

Bisindolylmaleimide VII

96% (TLC), solid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H27N5O2
CAS Number:
Molecular Weight:
453.54
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352111
MDL number:
Assay:
96% (TLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

96% (TLC)

form

solid

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

O=C1NC(=O)C(c2cn(CCCN3CCNCC3)c4ccccc24)=C1c5c[nH]c6ccccc56

InChI

1S/C27H27N5O2/c33-26-24(20-16-29-22-8-3-1-6-18(20)22)25(27(34)30-26)21-17-32(23-9-4-2-7-19(21)23)13-5-12-31-14-10-28-11-15-31/h1-4,6-9,16-17,28-29H,5,10-15H2,(H,30,33,34)

InChI key

VKLVUAHQOFDKLR-UHFFFAOYSA-N

Biochem/physiol Actions

Selective inhibitor of protein kinase C.

Disclaimer

Protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library