Skip to Content
Merck
All Photos(1)

Key Documents

B3640

Sigma-Aldrich

Anti-Biotin antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Biotin, Anti Biotin Antibody, Anti Biotin Antibody - Anti-Biotin antibody produced in goat, Anti-Biotin Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

immunoelectrophoresis: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is considered as an essential nutrient, though it is know to be secreted by some of the intestinal bacteria. It is found in liver and brain tissues in minute quantities.
Biotin:avidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay, the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.
Specificity and reactivity are determined by Ouchterlony Double Diffusion and immunoelectrophoresis versus the biotin-carrier protein complex. No reactivity is observed versus the carrier protein.

Immunogen

Biotin-KLH conjugate

Application

Anti-Biotin antibody produced in goat has been used in multi-well plate assay, to identify desthiobiotinylating (analog of biotin) proteins. It has also been used in Colourimetric enzyme immunoassay.
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.
May be substituted for avidin to decrease non-specific binding.

Biochem/physiol Actions

Biotin serves as a cofactor for carboxylases, necessary for oxidative metabolism. Biotin absorption in small intestine is reduced on consumption of avidin rich food such as egg white. Thus, greatly decreasing its bioavailability, leading to egg white injury. Reduced levels of biotin is normally observed during pregnancy. Biotin deficiency might be congenital, with defective proteins involved in biotin homeostasis.

Physical form

Lyophilized from 0.01 M sodium phosphate, 15 mM sodium chloride, pH 7.2

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen R Thom et al.
The Journal of biological chemistry, 289(27), 18831-18845 (2014-05-29)
This investigation was to elucidate the mechanism for microparticle (MP) formation triggered by exposures to high pressure inert gases. Human neutrophils generate MPs at a threshold of ∼186 kilopascals with exposures of 30 min or more. Murine cells are similar
Marcus Braun et al.
Nature chemical biology, 13(12), 1245-1252 (2017-10-17)
Microtubule-crosslinking motor proteins, which slide antiparallel microtubules, are required for the remodeling of microtubule networks. Hitherto, all microtubule-crosslinking motors have been shown to slide microtubules at a constant velocity until no overlap remains between them, leading to the breakdown of
Valerie Siahaan et al.
Nature cell biology, 21(9), 1086-1092 (2019-09-05)
Tau is an intrinsically disordered protein, which diffuses on microtubules1. In neurodegenerative diseases, collectively termed tauopathies, malfunction of tau and its detachment from axonal microtubules are correlated with axonal degeneration2. Tau can protect microtubules from microtubule-degrading enzymes such as katanin3.
Chao Meng et al.
Molecular medicine reports, 22(5), 4213-4220 (2020-10-02)
Renal ischemia reperfusion injury (IRI) after surgery may promote acute lung injury (ALI) by inducing an inflammatory response. However, the underlying molecular mechanism is still unclear. Studies have reported that inhibitor of κB kinase (IKK)ε primarily regulates inflammation and cell proliferation. The present study
Zdenek Lansky et al.
Cell, 160(6), 1159-1168 (2015-03-10)
Cytoskeletal remodeling is essential to eukaryotic cell division and morphogenesis. The mechanical forces driving the restructuring are attributed to the action of molecular motors and the dynamics of cytoskeletal filaments, which both consume chemical energy. By contrast, non-enzymatic filament crosslinkers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service