Skip to Content
Merck
All Photos(2)

Key Documents

B3555

Sigma-Aldrich

BMP2, human

≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

BMP-2

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

mol wt

26 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

solubility

water: soluble 0.100 mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BMP2(650)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cellular responses to BMP-2 are mediated by the formation of hetero-oligomeric complexes of type I and type II serine/threonine kinase receptors. BMPs stimulate angiogenesis by inducing the production of VEGF-A by osteoblasts. Human, mouse, and rat BMP-2 demonstrate 100% homology.

Physical form

Lyophilized from a 0.2 μm filtered buffered solution containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to induce alkaline phosphatase production by ATDC5 chondrogenic cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaxuan Chen et al.
Journal of tissue engineering and regenerative medicine, 10(1), 40-51 (2013-06-21)
1α,25-Dihydroxyvitamin D3 [1α,25(OH)2D3] and bone morphogenetic protein-2 (BMP2) are both used to stimulate osteoblastic differentiation. 1α,25(OH)2D3 regulates osteoblasts through classical steroid hormone receptor mechanisms and through rapid responses that are mediated by two receptors, the traditional vitamin D receptor (VDR)
M Kawabata et al.
Cytokine & growth factor reviews, 9(1), 49-61 (1998-08-28)
Bone morphogenetic proteins (BMPs) are multifunctional cytokines, which are members of the transforming growth factor-beta (TGF-beta) superfamily. Activities of BMPs are extracellularly regulated by BMP-binding proteins, Noggin and Chordin. BMPs bind to two different types of serine-threonine kinase receptors, type
Eun-Cheol Lee et al.
International journal of molecular sciences, 21(16) (2020-08-17)
The role of dental pulp stem cells (DPSCs) in dental tissue regeneration is gaining attention because DPSCs can differentiate into odontoblasts and other specialized cell types. Epigenetic modification has been found to play an important role in cell differentiation and
Martine M L Deckers et al.
Endocrinology, 143(4), 1545-1553 (2002-03-19)
During bone formation and fracture healing there is a cross-talk between endothelial cells and osteoblasts. We previously showed that vascular endothelial growth factor A (VEGF-A) might be an important factor in this cross-talk, as osteoblast-like cells produce this angiogenic factor
Lindsay A Bonsignore et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(7), 979-987 (2015-02-14)
The most important factor contributing to short-term and long-term success of cementless total joint arthroplasties is osseointegration. Osseointegration leads to a direct structural and functional connection between living bone and the surface of an implant. Surface contaminants may remain on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service