Skip to Content

B3313

Bicyclol

≥98% (HPLC)

Synonym(s):

4, 4′-dimethoxy-2, 3, 2′, 3′-bis(methylenedioxy)-6-hydroxymethyl-6′-methoxy-carbonyl biphenyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18O9
CAS Number:
Molecular Weight:
390.34
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COC(=O)c1cc(OC)c2OCOc2c1-c3c(CO)cc(OC)c4OCOc34

InChI

1S/C19H18O9/c1-22-11-4-9(6-20)13(17-15(11)25-7-27-17)14-10(19(21)24-3)5-12(23-2)16-18(14)28-8-26-16/h4-5,20H,6-8H2,1-3H3

InChI key

KXMTXZACPVCDMH-UHFFFAOYSA-N

General description

Bicyclol has anti-inflammatory properties and enhances the prognosis of kidney transplantation. It is involved in the removal of reactive oxygen species and modulates cytokine secretion.

Biochem/physiol Actions

Bicyclol is a anti-hepatitis drug, used in China for chronic hepatitis B and presumably C.
Bicyclol is a anti-hepatitis drug, used in China for chronic hepatitis B and presumably C. The drug appears to protect against liver injury in animals. Recently it was reported that bicyclol induce the expression of hepatic HSP27 and HSP70, and thus consequently inhibits the transcription factor NF-?B-mediated apoptosis and necrosis in liver tissue. It appears that bicyclol induces hepatic HSP27 and HSP70 expression through activating transcription of HSPs genes.


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library