Skip to Content
Merck

Skip To

B20221

Collagenase-DNase I blend

Tissue dissociation blend, lyophilized powder, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
(Collagenase (FALGPA assay) 1.0-5.0 units/ml in 10 ml HBSS (#H6648)), (Deoxyribonuclease I activity: 200-2000 Kunitz units/ml in 10 ml HBSS (#H6648))
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized powder

specific activity

(Collagenase (FALGPA assay) 1.0-5.0 units/ml in 10 ml HBSS (#H6648)), (Deoxyribonuclease I activity: 200-2000 Kunitz units/ml in 10 ml HBSS (#H6648))

storage temp.

-10 to -25°C

General description

The collagenase-DNase I blend is a tissue dissociation enzyme cocktail for cell isolation. The tissue dissociation process consists in the release of functional and viable cells from animal tissue, with minimal impact on the cell’s viability. The main reagent used for the tissue dissociation process is Collagenase. Collagenases are enzymes that break down the native collagen that holds animal tissues together. The second enzyme found in our blend is Deoxyribonuclease I (DNase I). DNase I, is an endonuclease of the DNase family, with a specific role for the degradation of DNA. DNase I is one of the most well characterized endonucleases of mammalian origin. During tissue dissociation, parts of the cells are lysed resulting in a release of DNA. Monomolecular DNA may cause clumping of cells, so addition of DNase I to our blend leads to a degradation of this extracellular DNA, thereby avoiding the loss of cells from undesired clumping.

Biochem/physiol Actions

Collagenase: Effective release of cells from tissue requires the action of collagenase enzymes. Collagenase is activated by four-gram atom calcium (Ca2+) per mole enzyme. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL are used for digestion. The duration of reaction varies from 15 minutes to several hours and yields satisfactory cell dissociation without causing too much cell death. DNase: DNase I is an endonuclease that acts on phosphodiester bonds adjacent to pyrimidines to produce polynucleotides with terminal 5′-phosphates. In the presence of Mg2+, DNAse I cleaves each strand of DNA independently and the cleavage sites are random. Both DNA strands are cleaved at approximately the same site in the presence of Mn2+. The pH optimum is found to be between 7 and 8. Divalent cations such as Mn2+, Ca2+, Co2+, and Zn2+ are activators of the enzyme. A concentration of 5 mM Ca2+ stabilizes the enzyme against proteolytic digestion. DNAse I from bovine pancreas consists of four chromatographically distinguishable components, A, B, C, and D, with their molar ratios being 4:1:1. Only minor amounts of D are found. 2-Mercaptoethanol, chelators, sodium dodecyl sulfate (SDS) and actin are known to inhibit the enzyme activity.

Preparation Note

For a working concentration of: Collagenase of 1 mg/ml and Deoxyribonuclease I of 0.1 mg/ml, reconstitute the lyophilized powder with 10 ml of Hank’s Balanced Salt solution with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate (#H6648)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B20222SCR139SCR140
specific activity

(Collagenase (FALGPA assay) 1.0-5.0 units/ml in 10 ml HBSS (#H6648)), (Deoxyribonuclease I activity: 200-2000 Kunitz units/ml in 10 ml HBSS (#H6648))

specific activity

(Collagenase activity : 1-5 units/ml in 10 ml HBSS (#H6648; FALGPA assay)), (Hyaluronidase activity : 80-400 units/ml in 10 ml HBSS (#H6648; Hyaluronidase assay))

specific activity

-

specific activity

-

form

lyophilized powder

form

lyophilized powder

form

-

form

-

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters