Skip to Content
Merck

B2013

Sodium 1-butanesulfonate

anionic detergent

Synonym(s):

1-Butanesulfonic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)3SO3Na
CAS Number:
Molecular Weight:
160.17
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
219-201-1
Beilstein/REAXYS Number:
3716662
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

mol wt

160.17 g/mol

mp

>300 °C (lit.)

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless

SMILES string

[Na+].CCCCS([O-])(=O)=O

InChI

1S/C4H10O3S.Na/c1-2-3-4-8(5,6)7;/h2-4H2,1H3,(H,5,6,7);/q;+1/p-1

InChI key

XQCHMGAOAWZUPI-UHFFFAOYSA-M

General description

Sodium 1-butanesulfonate is an anionic detergent.

Application

Ion-associating reagent for HPLC, including analyses of peptides and proteins.

Biochem/physiol Actions

Sodium 1-butanesulfonate can be used in the preparation of mobile phase for high performance liquid chromatography-inductively coupled plasma-mass spectrometry (HPLC-ICP-MS).


Still not finding the right product?

Explore all of our products under Sodium 1-butanesulfonate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hancock, W.S., et al.
Journal of Chromatography A, 11, 291-291 (1978)
Jian Zheng et al.
Analytical and bioanalytical chemistry, 374(2), 348-353 (2002-09-27)
The stability of five selenium compounds, selenate, Se(VI), selenourea, SeUr, trimethylselenonium ion, TMSe(+), selenomethionine, SeMet, and selenoethionine, SeEt, at concentrations from 30-60 micro g L(-1) in a pooled human urine, stored in dark at -20 degrees C, 4 degrees C
Bidlingmeyer, B.A., et al.
Advances in Chromatography, 14, 435-435 (1979)



Global Trade Item Number

SKUGTIN
B2013-25G04061835542833