Skip to Content
Merck
All Photos(3)

Documents

B1520

Sigma-Aldrich

Anti-Bovine Albumin antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Albumin, bovine serum (BSA)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

bovine

technique(s)

indirect ELISA: 1:200,000
quantitative precipitin assay: 3.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Serum albumin in humans is encoded by the gene mapped to human chromosome 4q13.3. Serum albumin is one of the abundant proteins in the blood and has a molecular weight of 66 kDa.
Anti-Bovine Serum Albumin (BSA) was developed in rabbit using purified BSA from Cohn Fraction V as the immunogen. The antiserum has been treated to remove lipoproteins. Strong reactivity with BSA has been determined by immunoelectrophoresis (IEP).

Application

Anti-Bovine Albumin antibody produced in rabbit may be used for ELISA at a working dilution of 1:200,000. The antibody was used for in vitro labeling of albumin present in mammary tissue extract of lactating cow.
It has also been used for the detection of albumin in nanoparticle using scanning transmission electron microscope.

Biochem/physiol Actions

Albumin is the most abundant circulating proteins present in the serum that binds and transports long-chain fatty acid anions, detoxifies unconjugated bilirubin and transports thyroid and steroid hormones.
Human serum albumin as a transporter, is considered as drug delivery vehicle for treating human diseases. Post-translational modifications like glycation and cysteinylation affects the binding functionality of serum albumin. High albumin protein diet has influence on hyperalbuminemia. Low levels of albumin leads to hypoalbuminemia, associated with decreased production and distribution. Hypoalbuminemia is a risk factor for the development of cardiovascular disease.

Physical form

Rabbit Anti-BSA is provided as a liquid containing 15 mM sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modifications of human serum albumin and their binding effect.
Lee P and Wu X
Current Pharmaceutical Design, 21(14), 1862-1865 (2015)
Xinmei Zhu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(5), 2483-2495 (2014-07-20)
Recent studies showed loss of CD36 or scavenger receptor-AI/II (SR-A) does not ameliorate atherosclerosis in a hyperlipidemic mouse model, suggesting receptors other than CD36 and SR-A may also contribute to atherosclerosis. In this report, we show that apolipoprotein E (apoE)-CD16
Albumin in health and disease: causes and treatment of hypoalbuminemia.
Throop JL, et al.
Compendium (Yardley, PA) (2004)
A R Ribeiro et al.
Scientific reports, 6, 23615-23615 (2016-03-30)
Dentistry and orthopedics are undergoing a revolution in order to provide more reliable, comfortable and long-lasting implants to patients. Titanium (Ti) and titanium alloys have been used in dental implants and total hip arthroplasty due to their excellent biocompatibility. However
Unraveling the mysteries of serum albumin?more than just a serum protein.
Merlot AM, et al.
Frontiers in Physiology, 5, 299-299 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service