Skip to Content
Merck
All Photos(1)

Key Documents

B1152

Sigma-Aldrich

Betamethasone 17,21-dipropionate

Synonym(s):

9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97.0%

form

powder

originator

Schering Plough

SMILES string

[H][C@@]12C[C@H](C)[C@](OC(=O)CCl)(C(=O)COC(=O)CC)[C@@]1(C)C[C@H](O)C3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C27H34ClFO7/c1-5-22(33)35-14-21(32)27(36-23(34)13-28)15(2)10-19-18-7-6-16-11-17(30)8-9-24(16,3)26(18,29)20(31)12-25(19,27)4/h8-9,11,15,18-20,31H,5-7,10,12-14H2,1-4H3/t15-,18-,19-,20-,24-,25-,26?,27-/m0/s1

InChI key

UOCNBCVPZWLUKX-HOBFILNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betamethasone dipropionate is a class III intermediate potency synthetic fluorinated corticosteroid, which is used to treat psoriasis vulgaris. It is also used to treat seborrheic dermatitis and chronic hand eczema.

Biochem/physiol Actions

Betamethasone 17,21-dipropionate is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Features and Benefits

This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Early onset of action and efficacy of a combination of calcipotriene and betamethasone dipropionate in the treatment of psoriasis
Papp KA, et al.
Journal of the American Academy of Dermatology, 48(1), 48-54 (2003)
Comparison of cyclosporine and topical betamethasone-17, 21-dipropionate in the treatment of severe chronic hand eczema.
Granlund H, et al.
Acta Dermato-Venereologica, 76(5), 371-376 (1996)
Seborrheic dermatitis
Gupta AK and Bluhm R
Journal of the European Academy of Dermatology and Venereology : JEADV, 18(1), 13-26 (2004)
Srinath Reddy et al.
Acta paediatrica (Oslo, Norway : 1992), 101(3), e130-e133 (2011-11-23)
Phimosis is a common paediatric urological disorder and often necessitates circumcision. We prospectively evaluated local steroid therapy (LST) as the first choice therapy for such children. Two hundred and sixty symptomatic boys up to 15 years of age (mean 34
Naresh Punwani et al.
Journal of the American Academy of Dermatology, 67(4), 658-664 (2012-01-28)
Janus-associated kinases (JAKs) are involved in signal transduction from a variety of cytokines implicated in the pathogenesis of psoriasis, including interleukin (IL)-12, IL-23, and interferon-γ. INCB018424, a small molecule inhibitor of JAK1 and JAK2, inhibits cytokine-induced JAK/signal transducers and activators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service