Skip to Content
Merck
All Photos(1)

Key Documents

B0519

Sigma-Aldrich

Biotin−Agarose

PBS suspension

Synonym(s):

Vitamin H Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

PBS suspension

Quality Level

extent of labeling

≥30 mg avidin per mL packed resin
2-6 μmol per mL

matrix

4% beaded agarose

matrix activation

epoxy

matrix attachment

carboxy (amide linkage); resin remains relatively uncharged over wide pH range

matrix spacer

13 atoms

application(s)

life science and biopharma

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Biotin contains a high affinity for binding to avidin or streptavidin. It has been widely used in biochemistry and molecular biology for detecting and labeling proteins.

Physical form

Suspension in phosphate buffered saline containing 0.02% sodium azide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biotinylated CdSe/ZnSe nanocrystals for specific fluorescent labeling.
Charvet, N., et al.
Journal of Materials Chemistry, 14, 2638-2642 (2004)
Dietmar Eberbeck et al.
Journal of nanobiotechnology, 6, 4-4 (2008-03-13)
The binding reaction of the biomolecules streptavidin and anti-biotin antibody, both labelled by magnetic nanoparticles (MNP), to biotin coated on agarose beads, was quantified by magnetorelaxometry (MRX). Highly sensitive SQUID-based MRX revealed the immobilization of the MNP caused by the
Emrah Celik et al.
Journal of molecular recognition : JMR, 25(1), 53-56 (2012-01-04)
Sample-probe contact duration (dwell time) and loading force are two important parameters for the atomic force microscopy (AFM) force spectroscopy measurements of ligand-receptor interaction. A prolonged contact time may be required to initiate ligand-receptor binding as a result of slow
P A Smith et al.
Nucleic acids research, 26(6), 1414-1420 (1998-04-29)
The high affinity binding interaction of biotin to avidin or streptavidin has been used widely in biochemistry and molecular biology, often in sensitive protein detection or protein capture applications. However, in vitro chemical techniques for protein biotinylation are not always
K J Airenne et al.
Gene, 144(1), 75-80 (1994-06-24)
A recombinant avidin (re-Avd), containing amino acids (aa) 1-123 of the native chicken egg-white Avd, was produced in Escherichia coli. When cells were grown at 37 degrees C production was over 1 microgram/ml, due to altering the codon preference of

Related Content

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service