Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polycarbonate , round bottle, screw top cap (Sealing), translucent bottle, translucent polycarbonate , translucent polypropylene screw closure
Parameter:
27500 × g max. RCF
Feature:
RCF 27,500 × g, with cap (sealing)
Packaging:
pack of 4
material
polycarbonate , round bottle, screw top cap (Sealing), translucent bottle, translucent polycarbonate , translucent polypropylene screw closure
joint
Joints threaded neck (29)
feature
RCF 27,500 × g, with cap (sealing)
packaging
pack of 4
manufacturer/tradename
Nalgene® 3122-0250
parameter
27500 × g max. RCF
bottle diam. × L
61.77 mm × 125.2 mm
nominal capacity
250 mL
Features and Benefits
- Nalgene® centrifuge bottles are used for large volume cell harvesting, pelleting, protein purification, and low-speed centrifugation of biological/chemical samples.
- Translucent - for better sample clarity.
- Spherical bottom design assists pellet formation and retrieval.
- Strong with excellent mechanical properties.
- Autoclavable - The bottles can withstand temperatures up to +121°C.
- For proper performance, bottles must be filled to at least 80% of the total capacity.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge bottles, Style 3122
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.