Skip to Content
Merck

AXYHS200

Axygen PlateMax heat sealing film for low temperature compound storage and PCR

peelable, 100 films/unit, 5 units/case

Synonym(s):

sealing mat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41122410
Manufacturer/tradename:
Axygen Inc HS-200, Corning
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

form

film

manufacturer/tradename

Axygen Inc HS-200, Corning

General description

  • Axygen® PlateMax® heat sealing films offer high-temperature resistance.
  • Made from polyester/aluminum
  • Aluminum material is for light-sensitive samples and polyester-based sealing films are ideal for short-term storage/incubation.
  • Compatible with the Axygen® PlateMax® Semi-Automated Plate Sealer (HS-1120, HS-1230)
  • Suitable for low-temperature compound storage, short-term room temperature compound storage and PCR.

Legal Information

Axygen is a registered trademark of Corning, Inc.
PlateMax is a registered trademark of Axygen, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library