AXYFXF165WBRS
Pipette filter tips for Biomek® FX, 165 μL, clear, wide bore, racked, sterile
pkg of 5x960tips/cs
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
volume:
165 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc FXF-165-WB-R-S
Corning
Corning
Recommended Products
manufacturer/tradename
Axygen Inc FXF-165-WB-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
165 μL
volume
165 μL
color
clear
Other Notes
Browse all pipette tips
Legal Information
Biomek is a registered trademark of Beckman Instruments, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service