Skip to Content
Merck
All Photos(1)

Key Documents

AV46141

Sigma-Aldrich

Anti-PPIF antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CYP3, Anti-Cyp-D, Anti-FLJ90798, Anti-MGC117207, Anti-Peptidylprolyl isomerase F (cyclophilin F)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

19 kDa

species reactivity

bovine, rat, dog, horse, rabbit, human, guinea pig, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPIF(10105)

Immunogen

Synthetic peptide directed towards the middle region of human PPIF

Application

Anti-PPIF antibody produced in rabbit is suitable for western blotting at a concentration of 5μg/ml.

Biochem/physiol Actions

PPIF (peptidylprolyl isomerase F) also referred to as CypD or cyclophilin F belongs to peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases are highly-conserved cytoplasmic enzymes that accelerate protein folding. PPIF is a part of the mitochondrial permeability transition pore (mPTP) in the inner mitochondrial membrane. Hence, it is anticipated that its association with the mPTP masks the binding site for inhibiting inorganic phosphate (Pi) as well as enhances the open possibility of mPTP that results in apoptosis or necrosis.

Sequence

Synthetic peptide located within the following region: GSTFHRVIPSFMCQAGDFTNHNGTGGKSIYGSRFPDENFTLKHVGPGVLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelina V Vaseva et al.
Cell, 149(7), 1536-1548 (2012-06-26)
Ischemia-associated oxidative damage leading to necrosis is a major cause of catastrophic tissue loss, and elucidating its signaling mechanism is therefore of paramount importance. p53 is a central stress sensor responding to multiple insults, including oxidative stress to orchestrate apoptotic
Roman A Eliseev et al.
The Journal of biological chemistry, 284(15), 9692-9699 (2009-02-21)
Cyclophilin D (CypD) is a mitochondrial immunophilin and a key positive regulator of the mitochondrial permeability transition (MPT). Several reports have shown that CypD is overexpressed in various tumors, where it has an anti-apoptotic effect. Because the MPT is a
Yuan He et al.
Investigative ophthalmology & visual science, 49(11), 4912-4922 (2008-07-11)
Disruption in intracellular calcium ion (Ca(2+)) homeostasis has major effects on health. Persistent Ca(2+) overload induces mitochondrial permeability transition pore (MPTP) opening, which prompts mitochondrial release of calcium (mCICR) and reactive oxygen species (ROS) into the cytosol which, in turn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service