Skip to Content
Merck

AV46090

Anti-PEX3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Peroxisomal biogenesis factor 3, Anti-TRG18

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

horse, human, mouse, guinea pig, dog, rabbit, rat, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PEX3(8504)

General description

Peroxisomal biogenesis factor 3 (PEX3, TRG18), a PEX19 docking protin, and PEX19 have key roles in the biogenesis of peroxisomes. Membrane-anchored PEX3 functions as a PEX19 receptor, wherein PEX19 recognizes newly synthesized peroxisomal membrane proteins (PMP) and recruits them to the peroxisomal membrane.

Immunogen

Synthetic peptide directed towards the N terminal region of human PEX3

Application

Anti-PEX3 polyclonal antibody is used to tag peroxisomal biogenesis factor 3 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of peroxisomal biogenesis factor 3 in peroxisome biosynthesis.

Biochem/physiol Actions

Anti-PEX3 polyclonal antibody reacts with bovine, human, mouse, rat, canine, zebrafish, and chicken peroxisomal biogenesis factor 3 proteins.
PEX3 is involved in peroxisome biosynthesis and integrity. It assembles membrane vesicles before the matrix proteins are translocated. As a docking factor for PEX19, it is necessary for the import of peroxisomal membrane proteins in the peroxisomes.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: KYGQKKIREIQEREAAEYIAQARRQYHFESNQRTCNMTVLSMLPTLREAL

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library