Skip to Content
Merck
All Photos(2)

Key Documents

AV36993

Sigma-Aldrich

Anti-TFAM (AB3) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AI661103, Anti-Hmgts, Anti-MtTFA, Anti-Transcription factor A, mitochondrial, Anti-TsHMG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

antibody form

affinity isolated antibody

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

27 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Tfam(21780)

Related Categories

Immunogen

Synthetic peptide directed towards the C terminal region of mouse Tfam

Biochem/physiol Actions

TFAM a mitochondrial transcription factor that is a key activator of mitochondrial transcription as well as a participant in mitochondrial genome replication. Studies in mice have demonstrated that this protein is required to regulate the mitochondrial genome copy number and is essential for embryonic development. A mouse model for Kearns Sayre syndrome was produced when expression of the gene was eliminated by targeted disruption in heart and muscle cells.

Sequence

Synthetic peptide located within the following region: FQEAKDDSAQGKLKLVNEAWKNLSPEEKQAYIQLAKDDRIRYDNEMKSWE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partial Inhibition of Complex I Restores Mitochondrial Morphology and Mitochondria-ER Communication in Hippocampus of APP/PS1 Mice.
Panes, et al.
Cells, 12 (2023)
Andrea Stojakovic et al.
Communications biology, 4(1), 61-61 (2021-01-10)
Alzheimer's Disease (AD) is a devastating neurodegenerative disorder without a cure. Here we show that mitochondrial respiratory chain complex I is an important small molecule druggable target in AD. Partial inhibition of complex I triggers the AMP-activated protein kinase-dependent signaling
Jonathan J Herrera et al.
Physiological reports, 12(9), e15997-e15997 (2024-05-03)
Voluntary or forced exercise training in mice is used to assess functional capacity as well as potential disease-modifying effects of exercise over a range of cardiovascular disease phenotypes. Compared to voluntary wheel running, forced exercise training enables precise control of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service