Skip to Content
Merck
All Photos(1)

Key Documents

AV35855

Sigma-Aldrich

Anti-ZP3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Zona pellucida glycoprotein 3 (sperm receptor)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

rabbit, bovine, horse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZP3(22932)

Immunogen

Synthetic peptide directed towards the C terminal region of human ZP3

Biochem/physiol Actions

ZP3 protein is a structural component of zona pellucida, an extracellular matrix that surrounds the oocyte and the early embryo. ZP3 mediates the binding and induction of the sperm acrosome reaction.

Sequence

Synthetic peptide located within the following region: NKGDCGTPSHSRRQPHVMSQWSRSASRNRRHVTEEADVTVGATDLPGQEW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pankaj Bansal et al.
The Indian journal of medical research, 130(1), 37-43 (2009-08-25)
An inability or decreased ability of spermatozoa to bind to the zona pellucida (ZP), an extracellular glycoproteinaceous matrix surrounding egg, is one of the plausible causes of idiopathic infertility. It will be clinically useful to distinguish this condition from other
Lei-Lei Gao et al.
Scientific reports, 7, 41272-41272 (2017-02-02)
ZP3 is a principal component of the zona pellucida (ZP) of mammalian oocytes and is essential for normal fertility, and knockout of ZP3 causes complete infertility. ZP3 promotes fertilization by recognizing sperm binding and activating the acrosome reaction; however, additional
Pankaj Bansal et al.
Biology of reproduction, 81(1), 7-15 (2009-02-28)
Zona pellucida glycoprotein 3 (ZP3) has been ascribed as a putative primary sperm receptor during fertilization in humans. Herein, attempts have been made to delineate the functional domain of human ZP3. ZP3 has been cloned and expressed in a baculovirus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service