Skip to Content
Merck
All Photos(8)

Key Documents

AMAB91153

Sigma-Aldrich

Anti-GAPDH Antibody

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, mouse monoclonal, CL3266

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Monoclonal Anti-GAPDH antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3266, purified immunoglobulin, buffered aqueous glycerol solution

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CL3266, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 1 μg/mL
immunofluorescence: 2-10 μg/mL (Fixation/Permeabilization: PFA/Triton X-100)
immunohistochemistry: 1:50000-1:100000

isotype

IgG2a

immunogen sequence

TVKAENGKLVINGNPITIFQERDPSKIKWGDAGAEYVVESTGVFTTMEKAGAHLQGGAKRVIIS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)

Immunogen

glyceraldehyde-3-phosphate dehydrogenase

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84470

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Xiong et al.
Oncology reports, 43(5), 1503-1515 (2020-04-24)
Long non‑coding RNA (lncRNA) small nucleolar RNA host gene 1 (SNHG1) was reported to be a critical regulator of tumorigenesis and is frequently deregulated in several cancer types. However, the exact mechanism by which SNHG1 contributes to breast cancer progression has not
Giulia Rizzo et al.
Nutrients, 15(7) (2023-04-14)
Background: Pomegranate (Punica granatum) can be used to prepare a bioactive extract exerting anti-inflammatory activities. Clinical studies demonstrated an improvement in clinical response in inflammatory bowel disease (IBD) patients when pomegranate extract (PG) was taken as a complement to standard
Xiaodan Chong et al.
Oncology letters, 25(1), 5-5 (2022-11-25)
Zinc finger DHHC-type palmitoyltransferase 9 (ZDHHC9) has been reported to play an important role in the occurrence and development of several types of cancer. However, its effects on colon cancer growth remain unclear. Using Gene Expression Profiling Interactive Analysis and
Ling Long et al.
Annals of translational medicine, 8(15), 927-927 (2020-09-22)
Neural stem cells (NSCs) are undifferentiated precursor cells that have the ability to self-renew and proliferate and have the capacity to become either glia (oligodendrocytes and astrocytes) or neurons. NSCs can act as beneficial adjuncts for many neurological disorders, such
Ou Li et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 8510-8523 (2018-11-26)
BACKGROUND Long non-coding RNAs (lncRNAs) have been implicated in various human cancer types. However, the underlying mechanisms involved in hepatocellular carcinoma (HCC) progression remain poorly understood. MATERIAL AND METHODS In this study, lncRNA array was used to identify HCC related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service