Skip to Content
Merck

Skip To

AF10

Aflatoxin Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
41100000
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

storage temp.

2-8°C

General description

Aflatoxin is the most common food toxin that is harmful to human and animal health. The most frequent aflatoxins are B1, B2, G1, and G2, which can affect the body through respiratory, mucosal, or cutaneous routes, causing an excessive inflammatory response. Aflatoxin can infect crops during their growing stages or even after they are harvested. It mainly targets the liver and can impair the effectiveness of immunization in children, increasing the risk of infection. Aflatoxin detection and quantification in food and feed is a critical part of food and feed safety concerns. The absorption and emission spectra of aflatoxins are commonly used to detect and identify them, with the highest absorbance occurring at 360 nm. Aflatoxin is frequently degraded, mitigated, and managed using a variety of treatments, including chemical, physical, and biological approaches.[1]
Aflatoxins are hepatotoxic metabolites. They are produced by molds, such as, Aspergillus flavus and Aspergillus parasiticus. They are naturally found poisonous carcinogens and the most widely known mycotoxins.

Packaging

Four aflatoxins from Aspergillus flavus

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9441A6655A6428
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • Aflatoxin B1 10 mg
    SDS

  • Aflatoxin B2 10 mg
    SDS

  • Aflatoxin G1 10 mg
    SDS

  • Aflatoxin G2 10 mg
    SDS

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is this instant kit? How many test possible in this kit?

    1 answer
    1. This product is simply a set of four Aflatoxin products, types B1, B2, G1, and G2, at a quantity of 10 mg each. It would be up to the end user to determine the specific assay protocol. Please see the links below to review the product information for each of the components in the set.
      Aflatoxin B1
      https://www.sigmaaldrich.com/product/sigma/a6636
      Aflatoxin B2
      https://www.sigmaaldrich.com/product/sigma/a9887
      Aflatoxin G1
      https://www.sigmaaldrich.com/product/sigma/a0138
      Aflatoxin G2
      https://www.sigmaaldrich.com/product/sigma/a0263

      Helpful?

Reviews

No rating value

Active Filters