Skip to Content
Merck
All Photos(1)

Key Documents

A9667

Sigma-Aldrich

Anti-Human IgE (ε-chain specific)−Peroxidase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgE (ε-chain specific)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:2,000
dot blot: 1:10,000 (chemiluminescent)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgEs are glycoprotein antibodies that regulate immunological responses to allergies and infections. These immunoglobulins have been implicated in mediating Type I hypersensitivity reactions . Elevated IgE levels have been associated with hyper IgE syndrome and allergic asthma. Anti-Human IgE ε-chain specific-Peroxidase antibody binds to human IgE as against human IgG, and Bence Jones κ and λ myeloma proteins.
Immunoglobulin E (IgE) belongs to the immunoglobulin family and consists of epsilon(ε) heavy chain in the constant (C) region. IgE has a monomeric structure with an extra domain in the constant region.

Immunogen

IgE purified from human myeloma serum

Application

Anti-Human IgE (ε-chain specific)-Peroxidase antibody produced in goat has been used in IgE immunoblotting and in western blotting to analyse IgE binding efficiency.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of different thermal processing methods on the structure and allergenicity of peanut allergen Ara h 1
Tian Y, et al.
Food Science & Nutrition, 6(6), 1706-1714 (2018)
Marília Porto Oliveira Nunes et al.
PloS one, 14(4), e0214745-e0214745 (2019-04-18)
Given the growing incidence and prevalence of life-threatening food allergies, health concerns have raised new perspectives for in vivo and in vitro diagnostic methodologies, pointing to saliva as a promising material, already used to diagnose other pathologies. Based on the
Y Shahali et al.
Allergy, 64(12), 1773-1779 (2009-07-25)
The allergenic characteristics of pollen and their levels of expression may vary depending on the plant species, the degree of maturation and the influence of environmental factors such as climate and atmospheric pollution. The objective of this survey was the
Amita Misra et al.
GM crops & food, 3(4), 273-282 (2012-06-30)
Genetically modified (GM) mustard line (V4) with increased carotenoid content was compared with native mustard to find the difference in allergenic potential, if any. Simulated gastric fluid (SGF) digestibility of crude protein extract from GM as well as its native
Paul Neybecker et al.
Stem cell research & therapy, 9(1), 329-329 (2018-11-30)
Mesenchymal stem cells (MSCs) are found in synovial fluid (SF) and can easily be harvested during arthrocentesis or arthroscopy. However, SF-MSC characterization and chondrogenicity in collagen sponges have been poorly documented as well as their hypothetical in vivo chondroprotective properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service