Skip to Content
Merck

A9607

Anti-ATP13A2 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ATPase type 13A2, Anti-PARK9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~129 kDa

species reactivity

mouse, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using mouse brain extract (S1 fraction), western blot: 2-4 μg/mL using extract of HEk-293T cells expressing human ATP13A2

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP13A2(23400)
mouse ... Atp13a2(74772)
rat ... Atp13a2(362645)

General description

ATP13A2 is a member of the P5 family of ATPases which function in the transport of inorganic cations.
ATPase type 13A2 (ATP13A2) is a neuronal P-type ATPAse. The gene is also referref as PARK9 (Parkinson′s disease 9).
Rabbit Anti-ATP13A2 antibody binds to human and mouse ATP13A2.

Application

Rabbit Anti-ATP13A2 antibody has been used for western blot assays.

Biochem/physiol Actions

Mutations in the gene encoding ATPase type 13A2 (ATP13A2) is implicated with a hereditary form of PD associated with dementia, known as Kufor-Rakeb syndrome (KRS). ATP13A2 shows elevated expression levels in brains of sporadic PD patients, suggesting a potential role in the more common forms of Parkinson′s disease.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

wgk

nwg

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library