Skip to Content
Merck
All Photos(2)

Documents

A9561

Sigma-Aldrich

5-(N,N-Hexamethylene)amiloride

Synonym(s):

HMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClN7O
CAS Number:
Molecular Weight:
311.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

N\C(N)=N\C(=O)c1nc(Cl)c(nc1N)N2CCCCCC2

InChI

1S/C12H18ClN7O/c13-8-10(20-5-3-1-2-4-6-20)18-9(14)7(17-8)11(21)19-12(15)16/h1-6H2,(H2,14,18)(H4,15,16,19,21)

InChI key

RQQJJXVETXFINY-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of Na+/H+ antiport.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Montserret et al.
The Journal of biological chemistry, 285(41), 31446-31461 (2010-07-30)
The small membrane protein p7 of hepatitis C virus forms oligomers and exhibits ion channel activity essential for virus infectivity. These viroporin features render p7 an attractive target for antiviral drug development. In this study, p7 from strain HCV-J (genotype
Lauren Wilson et al.
Virology, 353(2), 294-306 (2006-07-04)
All coronaviruses encode a small hydrophobic envelope (E) protein, which mediates viral assembly and morphogenesis by an unknown mechanism. We have previously shown that the E protein from Severe Acute Respiratory Syndrome coronavirus (SARS-CoV) forms cation-selective ion channels in planar
Gary D Ewart et al.
Antimicrobial agents and chemotherapy, 48(6), 2325-2330 (2004-05-25)
We report that the amiloride analogues 5-(N,N-hexamethylene)amiloride and 5-(N,N-dimethyl)amiloride inhibit, at micromolar concentrations, the replication of human immunodeficiency virus type 1 (HIV-1) in cultured human blood monocyte-derived macrophages. These compounds also inhibit the in vitro activities of the HIV-1 Vpu
Sandra Ribeiro de Morais et al.
Pharmacognosy magazine, 12(48), 262-270 (2016-11-22)
Lippia sidoides (Verbenaceae) is used in Brazilian folk medicine as an antiseptic, and it is usually applied topically on skin, mucous membranes, mouth, and throat, or used for vaginal washings. To analyze the chemical composition of the essential oil from
Gary D Ewart et al.
European biophysics journal : EBJ, 31(1), 26-35 (2002-06-06)
The Vpu protein of human immunodeficiency virus type 1 forms cation-selective ion channels and enhances the process of virion budding and release. Mutagenesis studies have shown that the N-terminal transmembrane domain primarily controls both of these activities. Here we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service