Skip to Content
Merck

A9543

Bovine Serum Albumin

heat shock fraction, low endotoxin, pH 7, ≥98%

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥98%
Form:
lyophilized powder
Technique(s):
ELISA: suitable, electrophoresis: suitable, western blot: suitable
Impurities:
≤1.0 EU/mg endotoxin
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥98%

form

lyophilized powder

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

ELISA: suitable, electrophoresis: suitable, western blot: suitable

impurities

≤1.0 EU/mg endotoxin

loss

≤5%

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

BT virus, none detected, VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Application

This A9543 BSA product has been used in various systems, including:
  • NRK-52E rat kidney epithelioid cells: Ochiai, H. et al., J. Virol., 67(11), 6811-6814 (1993)
  • Neonatal porcine pancreatic islets: Ellis, C.E. et al., Islets, 5(5), 216-225 (2013)
  • Mouse:
Lee, D. et al., Am. J. Physiol. Renal Physiol., 304(7), F1009-F1019 (2013)
Girbl, T. et al., Immunity, 49(6), 1062-1076.e6 (2018)
  • iPSC cell line: Martin, H.C. et al., Science, 362(6419), 1161-1164 (2018).

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Heat shock fractionated
  • Low endotoxin

Preparation Note

Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library