Skip to Content
Merck

A9524

DL-Arabinose

≥98% (GC)

Synonym(s):

(2R,3R,4R)-2,3,4,5-tetrahydroxypentanal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
205-699-8
Beilstein/REAXYS Number:
1723086
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

cell culture

Quality Segment

assay

≥98% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4-,5+/m0/s1

InChI key

PYMYPHUHKUWMLA-VAYJURFESA-N

Application

DL-Arabinose is a mixture of the diastereomers D-arabinose and L-arabinose used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
A9524-5G04061832387161
A9524-10G04061833402825
A9524-50G04061833402863