Skip to Content
Merck

Skip To

A9477

Aldosterone

≥95% (HPLC), powder

Synonym(s):

11β,21-Dihydroxy-3,20-dioxo-4-pregnen-18-al, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione, 18-Aldocorticosterone, Reichstein X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
EC Number:
200-139-9
MDL number:
Beilstein/REAXYS Number:
3224996
Assay:
≥95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Aldosterone, ≥95% (HPLC)

biological source

synthetic (organic)

Quality Segment

sterility

non-sterile

assay

≥95% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

chloroform: 20 mg/mL, clear to slightly hazy, colorless to light yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O

InChI

1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,11,14-17,19,22,25H,2-7,9-10H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1

InChI key

PQSUYGKTWSAVDQ-ZVIOFETBSA-N

Gene Information

General description

Aldosterone plays a significant role in the multi-factorial regulation of salt, potassium, blood pressure, and acid-base balance. It is also involved in maternal volume expansion during fetal development to support fetal perfusion and may contribute to the increased expression of placental growth factors. While its primary activity is associated with the kidney, aldosterone can also interact with mineralocorticoid receptors in other tissues such as the gastrointestinal tract, respiratory epithelium, myocardium, and vascular smooth muscle. Aldosterone antagonists or mineralocorticoid receptor antagonists (MRAs) can be beneficial for patients with various clinical conditions, including primary aldosteronism (PA), primary and resistant hypertension, heart failure (HF), and chronic kidney disease (CKD).

Application

Aldosterone has been used:
  • as a RPMI-1640 medium supplement for podocyte culture to test its pathogenicity in obesity-related glomerulopathy (ORG)
  • in combination with salt to induce renal injury in mice
  • to stimulate primary liver sinusoidal endothelial cells (LSECs) and in the generation of hyperaldosteronism model in mice

The d-isomer of aldosterone is considered to be the biologically active isomer.

Biochem/physiol Actions

Aldosterone is a biologically active aldosterone isomer; a mineralocorticoid produced by the adrenal cortex. It induces urinary excretion of K+ and renal reabsorption of Na+. It is produced from adrenocorticotropic hormone (ACTH) and is essential for maintaining sodium homeostasis in the distal tubules. Aldosterone binds to the mineralocorticoid receptor and may modulate vascular system and induce kidney damage. It contributes to the progression of chronic kidney disease(CKD). Aldosterone is part of the renin-angiotensin-aldosterone system (RAAS).

Other Notes

Exists as an equilibrium mixture of the aldehyde and the hemiacetal.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2505C2755E1001
form

powder

form

powder

form

powder

form

powder

assay

≥95% (HPLC)

assay

≥92%

assay

≥95%

assay

≥99% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

inhibition assay: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

solubility

chloroform: 20 mg/mL, clear to slightly hazy, colorless to light yellow

solubility

chloroform: 50 mg/mL, clear, colorless to faintly yellow

solubility

chloroform: methanol (1:1): 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

solubility

chloroform: methanol (1:1): 20 mg/mL, clear to slightly hazy, colorless to light yellow


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. Is Product A9477, Aldosterone, the d- or l- isomer?

    1 answer
    1. The A9477 is the d-Aldosterone (per our supplier), which is considered to be the biologically active isomer.

      Helpful?

  2. Is Product A9477, Aldosterone, tested for solubility?

    1 answer
    1. Yes, A9477 is tested for solubility using 20mg/ml in chloroform. For biological applications, you can solubilize aldosterone in ethanol to at least 1 mg/mL. Please refer to literature references for further solubility information.

      Helpful?

  3. What is the difference between Aldosterone Products A9477 and 05521?

    1 answer
    1. The main differences between A9477 and 05521 are the assays performed on the products. The assay results are lot specific and can be viewed on the Certificate of Analysis.

      Helpful?

  4. How do I make a stock solution of Product A9477, Aldosterone?

    1 answer
    1. To prepare a 100 μg/ml stock solution, add 5 ml of absolute ethanol to 5 mg, gently swirl to dissolve, then add 45 ml sterile medium. Freeze the stock in working aliquots, but avoid repeated freeze/thaw cycles.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters