Skip to Content
Merck
All Photos(2)

Key Documents

A9473

ART® self-sealing barrier pipette tips

ART XLP, volume range 5-100 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NB.01
maximum volume:
100 μL
volume range:
5-100 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
case of 4 packs
pack of 960 ea (Racked)
manufacturer/tradename:
Molecular BioProducts 2159P

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

case of 4 packs
pack of 960 ea (Racked)

manufacturer/tradename

Molecular BioProducts 2159P

technique(s)

PCR: suitable

maximum volume

100 μL

volume range

5-100 μL

Looking for similar products? Visit Product Comparison Guide

General description

Extended length for reaching the bottom of 12 × 75 tubes and vials

Application

ART® self-sealing barrier pipette tips have been used in PCR reactions and LC–MS/MS (liquid chromatography–mass spectrometry).

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Compatibility

Compatibility
Biohit Proline-Variable 5-50 μl/50-200 μl
Eppendorf 4810 10-100 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Research/Reference 10-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 Digital 5-40 μl
Mettler-Toledo VoluMate 10-100 μl
Nichiryo BenchMate 5-100 μl/20-200 μl
Pipetman P-100 10-100 μl
Rainin EDP 2 10-100 μl/20-250 μl
Rainin EDP Plus 10-100 μl/20-250 μl
Rainin Pipet Plus 10-100 μl/20-200 μl
Rainin Pipetman 10-100 μl/20-200 μl
Socorex/Wheaton Calibra/822 10-100 μl
Transferpette Multi-Channel 10-100 μl

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prolonged arthritis associated with sindbis-related (Pogosta) virus infection.
Laine M
Rheumatology (Oxford, England), 39, 1272-1274 (2000)
Nitrogenase genes in PCR and RT-PCR reagents: implications for studies of diversity of functional genes.
Zehr JP
Biotechniques, 35, 996-1002 (2003)
Quantitative profiling of endocannabinoids in lipoproteins by LC-MS/MS.
Bilgin M, et al.
Analytical and Bioanalytical Chemistry, 407, 5125-5131 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service