Skip to Content
Merck
All Photos(1)

Key Documents

A9459

Sigma-Aldrich

Apamin

synthetic, ≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C79H131N31O24S4
CAS Number:
Molecular Weight:
2027.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC(C)C[C@@H]1NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H]2CCC[N@H]2C(=O)[C@H](C)NC(=O)[C@H](CCCCN)NC(=O)[C@@H]3CSSC[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CSSC[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N3)NC1=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc4cnc[nH]4)C(N)=O)[C@@H](C)O

InChI

1S/C79H131N31O24S4/c1-35(2)26-49-70(127)107-51-31-136-135-30-41(81)63(120)105-50(28-57(84)114)71(128)108-53(73(130)99-42(12-7-8-22-80)64(121)96-38(5)77(134)110-25-11-15-54(110)75(132)102-47(18-21-58(115)116)69(126)109-59(39(6)111)76(133)95-37(4)62(119)104-49)33-138-137-32-52(106-66(123)44(14-10-24-92-79(88)89)98-65(122)43(13-9-23-91-78(86)87)97-61(118)36(3)94-72(51)129)74(131)101-45(16-19-55(82)112)67(124)100-46(17-20-56(83)113)68(125)103-48(60(85)117)27-40-29-90-34-93-40/h29,34-39,41-54,59,111H,7-28,30-33,80-81H2,1-6H3,(H2,82,112)(H2,83,113)(H2,84,114)(H2,85,117)(H,90,93)(H,94,129)(H,95,133)(H,96,121)(H,97,118)(H,98,122)(H,99,130)(H,100,124)(H,101,131)(H,102,132)(H,103,125)(H,104,119)(H,105,120)(H,106,123)(H,107,127)(H,108,128)(H,109,126)(H,115,116)(H4,86,87,91)(H4,88,89,92)/t36-,37-,38-,39+,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,59-/m0/s1

InChI key

YVIIHEKJCKCXOB-STYWVVQQSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Asn-Cys-Lys-Ala-Pro-Glu-Thr-Ala-Leu-Cys-Ala-Arg-Arg-Cys-Gln-Gln-His-NH2 [Disulfide Bridges: 1-11, 3-15]

General description

Apamin, a bee venom is a polypeptide, that is made of 18 amino acids with two disulfide bridges.

Application

Apamin has been used in microbiological assays for the determination of antimicrobial activity of synthetic apamin and as a standard for MALDI (matrix-assisted laser desorption/ionization matrix assisted laser desorption ionization-time of flight mass spectrometry (MALDI-TOF MS) and electrospray ionisation mass spectrometry (ESI-MS) analysis.

Biochem/physiol Actions

Apamin is a neurotoxin which can pass the blood-brain barrier. In human, it might cause peripheral nerve dysfunction, mainly seen after bee stings.
The only polypeptide neurotoxin that is known to pass the blood-brain barrier. Blocks ATP-type Ca+2-activated K+ channels.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio A Villalobos et al.
Cell reports, 39(3), 110699-110699 (2022-04-21)
We report that increasing inhibition from the basal ganglia (BG) to the superior colliculus (SC) through the substantia nigra pars reticulata (nigra) using in vivo optogenetic activation of GABAergic terminals in mice produces contralateral orienting movements. These movements are unexpected because
Apamin as a selective blocker of the calcium-dependent potassium channel in neuroblastoma cells: voltage-clamp and biochemical characterization of the toxin receptor
Hugues M, et al.
Proceedings of the National Academy of Sciences of the USA, 79(4), 1308-1312 (1982)
Zootoxins
Reproductive and Developmental Toxicology, 765-771 (2011)
Top-down sequencing of Apis dorsata apamin by MALDI-TOF MS and evidence of its inactivity against microorganisms
Baracchi D, et al.
Toxicon, 105-112 (2013)
Ramesh C Mishra et al.
Cardiovascular research, 97(2), 339-348 (2012-11-03)
Endothelial SK(Ca) and IK(Ca) channels play an important role in the regulation of vascular function and systemic blood pressure. Based on our previous findings that small molecule activators of SK(Ca) and IK(Ca) channels (i.e. NS309 and SKA-31) can inhibit myogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service