Skip to Content
Merck
All Photos(1)

Key Documents

A8679

Sigma-Aldrich

Anti-Aflatoxin B1 antibody produced in rabbit

fractionated antiserum, buffered aqueous solution

Synonym(s):

Anti-Aflatoxin B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

capture ELISA: 1:4,000 using aflatoxin B1-BSA

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aflatoxin B1 is the most abundant and significant member of the group, which includes B2, B2a, G1, G2a, M1, M2, P1, Q1, aflatoxicol I (natural isomer), aflatoxicol II (unnatural isomer), tetrahydrodeoxy aflatoxin B1, and the unstable reactive B1 (8,9)-epoxide.
Aflatoxins are highly carcinogenic.
Aflatoxins are highly toxic mycotoxins produced by the fungus Aspergillus. Aflatoxins are commonly found in various grains and peanut butter.
Mycotoxins, a group of chemically diverse secondary fungal metabolites, induce a variety of toxic responses in humans and animals when food and feed containing these compounds are ingested. Rapidly quantitating levels of mycotoxin contamination can help reduce exposure to these toxins.
The antibody reacts with the B1 aflatoxin and also recognizes aflatoxins G1, B2, and the KLH protein. No cross-reaction is observed with aflatoxins and B2a, G2, G2a, M1.

Specificity

Sensitivity range for this product is 0.005-20 ng/ml of aflatoxin B1.

Immunogen

aflatoxin B1-KLH.

Application

Rabbit Anti-Aflatoxin B1 antibody has been used for the development of electrochemical immunosensors. The antibody has also been used in designing novel sandwich immunoassays. The product can also be used for capture ELISA at a dilution of 1:1,000.

Biochem/physiol Actions

Aflatoxin B1 and its metabolites act as potent carcinogens, mutagens and teratogens. Aflatoxins have been implicated in human hepatocellular carcinoma, outbreaks of aflatoxicosis, Rey′s syndrome, chronic hepatitis and increased mortality from infection in animal husbandry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqiang Lai et al.
Biosensors & bioelectronics, 89(Pt 1), 645-651 (2016-01-05)
A new colorimetric immunosensing platform accompanying enzyme cascade amplification strategy was fabricated for quantitative screening of small-molecular mycotoxins (aflatoxin B1, AFB1 used in this case) coupling with enzyme-controlled dissolution of MnO2 nanoflakes. The visual colored assay was executed by high-efficient
Gilcelia J L S Barbieri et al.
Biotechnology progress, 39(3), e3334-e3334 (2023-02-17)
This article developed a novel electrochemical immunosensor for the specific detection of aflatoxin B1 (AFB1). Amino-functionalized iron oxide nanoparticles (Fe3 O4 -NH2 ) were synthesized. Fe3 O4 -NH2 were chemically bound on self-assembly monolayers (SAMs) of mercaptobenzoic acid (MBA). Finally
Jeong Moon et al.
Sensors (Basel, Switzerland), 18(2) (2018-02-22)
Aflatoxins (AFs) are highly toxic compounds that can cause both acute and chronic toxicity in humans. Aflatoxin B1 (AFB1) is considered the most toxic of AFs. Therefore, the rapid and on-site detection of AFB1 is critical for food safety management.
C G Caratsch et al.
Neuroscience letters, 111(3), 344-350 (1990-04-06)
The effects of calcitonin gene-related peptide (CGRP) on synaptic mechanisms were studied at the frog neuromuscular junction by using classical electrophysiological techniques. CGRP reduced the quantal content of evoked neurotransmitter release, as well as the sensitivity of postsynaptic nicotinic acetylcholine
An over review on effect of aflatoxin in animal husbandry
Talebi E, et al.
Asian journal of experimental biological sciences, 2(3), 754-757 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service