Skip to Content
Merck

A8671

Dodecyl arachidate

≥99%

Synonym(s):

Lauryl arachidate, Eicosanoic acid dodecyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H64O2
CAS Number:
Molecular Weight:
480.85
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

powder

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-15-16-17-18-19-20-21-22-24-26-28-30-32(33)34-31-29-27-25-23-14-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

GFFYHZLIDINABP-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library