Skip to Content
Merck
All Photos(1)

Key Documents

A8398

Sigma-Aldrich

7-Aminodesacetoxycephalosporanic acid

Synonym(s):

7-ADCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1=C(N2[C@H](SC1)[C@H](N)C2=O)C(O)=O

InChI

1S/C8H10N2O3S/c1-3-2-14-7-4(9)6(11)10(7)5(3)8(12)13/h4,7H,2,9H2,1H3,(H,12,13)/t4-,7-/m1/s1

InChI key

NVIAYEIXYQCDAN-CLZZGJSISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

7-Aminodesacetoxycephalosporanic acid is used in the synthesis of cephalosporins and for bioconversion studies .

Biochem/physiol Actions

7-ADCA is produced from penicillin G made by Penicillium chrysogenum involving several polluting chemical steps followed by enzymatic deacylation using penicillin acylase .

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda G Otten et al.
The Journal of biological chemistry, 277(44), 42121-42127 (2002-08-29)
Using directed evolution, we have selected an adipyl acylase enzyme that can be used for a one-step bioconversion of adipyl-7-aminodesacetoxycephalosporanic acid (adipyl-7-ADCA) to 7-ADCA, an important compound for the synthesis of semisynthetic cephalosporins. The starting point for the directed evolution
Mutational analysis of a key residue in the substrate specificity of a cephalosporin acylase.
Linda G. Otten, Charles F. Sio, et al.
Chembiochem, 6, 820-825 (2004)
P Travascio et al.
Biotechnology and bioengineering, 79(3), 334-346 (2002-07-13)
A new hydrophobic and catalytic membrane was prepared by immobilizing Penicillin G acylase (PGA, EC.3.5.1.11) from E. coli on a nylon membrane, chemically grafted with butylmethacrylate (BMA). Hexamethylenediamine (HMDA) and glutaraldehyde (Glu) were used as a spacer and coupling agent
Catharina Gerarda Petronella Henrica Schroën et al.
Biotechnology progress, 18(4), 745-751 (2002-08-03)
Adipyl-7-ADCA is a new source for 7-aminodeacetoxycephalosporanic acid (7-ADCA), one of the substrates for antibiotics synthesis. In this paper, a novel process for enzymatic 7-ADCA production is presented. The process consists of a reactor, a crystallization step, a membrane separation
P Travascio et al.
Biotechnology progress, 18(5), 975-985 (2002-10-05)
The effect of methanol on the kinetically controlled synthesis of cephalexin by free and immobilized penicillin G acylase (PGA) was investigated. Catalytic and hydrophobic membranes were obtained by chemical grafting, activation, and PGA immobilization on hydrophobic nylon supports. Butyl methacrylate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service